Learn More
Glass Expansion GazFit Union 8mm (PKT 2)
Supplier: Glass Expansion GAZ08U
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
GazFit Union 8mm (PKT 2). GazFit Union Connector for 8mm OD laser ablation adaptors and 8mm OD rigid walled tubing. Compatible with laser ablation adaptor 21-809-4140.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001