Learn More
Glass Expansion GazFit Connectors for 8mm OD torch side arms (PKT 2). Compatible with Glass Expansion Laser Ablation Adaptor 21-809-4140. Makes connection to 8mm OD side arms with soft walled tubing.
Supplier: Glass Expansion GAZ08
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
GazFit Connectors for 8mm OD torch side arms (PKT 2). Compatible with Glass Expansion Laser Ablation Adaptor 21-809-4140. Makes connection to 8mm OD side arms with soft walled tubing. For 4, 5 or 6mm OD connections, use GAZ-04, GAZ-5, GAZ-06, respectively.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001