Learn More
INDUSTRIAL PRODUCTS LIMITED LLC GAS CYLNDR SUPPORT BRKT W/CHN
SDP
Supplier:  INDUSTRIAL PRODUCTS LIMITED LLC 35256JR
Encompass
Cylinder support brackets provide a quick and easy solution for storing gas cylinders when space is at premium. Constructed from HDPE, the adjustable width design uses recessed set screws that lock the two halves in place.INDUSTRIAL PRODUCTS LIMITED LLC mfg# 35256JR
Catalog No. NC2293306