Learn More
Chemglass Life Sciences Funnel, Eco, 4" Top Diameter, to Fit 2 and 5 Gallon Justrite Containers
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG175922
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Funnel, Eco, 4" Top Diameter, to Fit 2 and 5 Gallon Justrite Containers. The 4” Eco Funnel is made of HDPE and has a 450mL fluid capacity. Features an easy to use lid, gasket and positive latch to close the lid and maintain an airtight seal. A filter is also built into the interior to reduce the loss of magnetic stir bars and other items when emptying the contents of a vessel into the funnel. The funnel is equipped with an 11” stem to reduce chemical evaporation of hazardous solvents in the lab.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.