Learn More
Wilmad Labglass FUNNEL ADD CYL GRAD 250ML
Supplier: Wilmad Labglass LG8271T104
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Funnel Addition, Graduated, Pressure Equalizing, PTFE Stopcock. Capacity: 250mL; Subdivisions: 5mL; Joint Size: 24/40; Stopcock Bore: 2. Addition funnel w/ double Scale graduations.
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov