Learn More
Medchemexpress LLC HY-N6719 1mg Medchemexpress, Fumonisin B1 CAS:116355-83-0 Purity:>98%
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Medchemexpress LLC HYN67191MG
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Medchemexpress, HY-N6719 1mg Fumonisin B1 CAS:116355-83-0 Fumonisin B1 is a mycotoxin produced from Fusarium moniliforme. Fumonisin B1 is a potent inhibitor of sphingosine N-acyltransferase (ceramide synthase) and disrupts de novo sphingolipid biosynthesis. Fumonisin B1 is the most abundant and toxic fumonisin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.