Learn More
Electron Microscopy Sciences Formvar/Carbon Film 5-6 nm/1 nm thick on Single 200 µm hole Copper Grid
Supplier: Electron Microscopy Sciences FCFGA200CUUA
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. Coated grids are optically checked followed by batch testing in the electron microscope. Packed in grid storage box. NOTE: All film is laid on the shiny side of the grid. Formvar is 5-6 nm thick with a 1 nm carbon coating.
Grid: Copper
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.