Learn More
Hampton Research Four Color Mounting Clay
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Hampton Research HR4326
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Adhesive, malleable, non-hardening clays have many uses in the crystallization lab. Useful for holding capillaries during crystal mounting as well as, stabilizing the capillary in the goniometer head. Useful for propping open the lids of crystallization plates. Great for color coding experiments, capillaries, and Linbro crystallization plates. Supplied as four 4 ounce bars of clay, each bar is individually wrapped. This Van Aken Claytoon Hot Set offers the colors Magenta, Rose, Neon Orange, and Yellow.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.