Learn More
TARGETMOL CHEMICALS INC FORMOTEROL FUMARATE 25MG
Supplier: TARGETMOL CHEMICALS INC T6505
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Formoterol fumarate (Eformoterol Hemifumarate) an effective specific and long-acting (beta)2-adrenoceptor agonist is utilized in the treatment of chronic obstructive pulmonary disease(COPD) and asthma. purity: 99%