Learn More
STA PHARMACEUTICAL US LLC WuXi TIDES Fmoc-D-Phe(3,4-DiCl)-OH | 177966-58-4, 100GR
Supplier: STA PHARMACEUTICAL US LLC STAAA10723100GM
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
WuXi TIDES Fmoc-D-Phe(3,4-DiCl)-OH, 177966-58-4 | (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(3,4-dichlorophenyl)propanoic acid | ClC=1C=CC(=CC1Cl)C[C@H](C(O)=O)NC(OCC2C3=CC=CC=C3C4=CC=CC=C42)=O | M.F. C24H19Cl2NO4, M.W. 456.32