Learn More
AVANTOR MASTERFLEX (VWR LLC) Hedland DDFXD-ANNA-NN DFX Doppler Flow Monitor, 0.15 To 30 FPS, No Input/Output
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX3261580
In-field linearization allows adaptation to a wide variety of applications. Accuracy is 2% full-scale over calibrated span. Nonintrusive, clamp-on transducers fit pipes from and above. Clamp-on sensor eliminates the need for in-line flanges, pipe fittings, strainers, or filters.