Learn More
AVANTOR MASTERFLEX (VWR LLC) Hedland H624-607 Flowmeter, spring-loaded in-line, for water, 1/2"" NPT(F), 1 to 7 gpm
Supplier: AVANTOR MASTERFLEX (VWR LLC) MFLX0323103
These economical flowmeters are built with a spring-retained piston for direct, accurate readings regardless of the position you mount them. Use them to monitor hydraulic systems or chemical processes; the meters also work well to check pump and control valve performance. Note: Polysulfone is not suited for direct contact with sunlight.
Considering limit switches? Use to provide an output signal at a setpoint anywhere between 0 to 100% of full-scale. Use to control small pumps, valves, or alarms or to interface with a PLC.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.