Learn More
Perkin Elmer US LLC UV Quartz SUPRASIL Rectangular Flow-Through Cell, Semi-Micro/Ultra-Micro with In- and Outlet Tubes - 390 µL, Qty. 1
Supplier: Perkin Elmer US LLC B0631084
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The UV Quartz SUPRASIL Flow-Through Cell has a light path of 10 mm, a volume of 390 µL, an aperture of 11 x 3.5 mm, and outside dimensions of 35 mm (H) x 12.5 mm (W) x 12.5 mm (D). Qty. 1