Learn More
Chemglass Life Sciences Flask, 2800mL, Fernbach, 70mm Screw Thread, Without Baffles, Vented Cap, Approx OD x Height (mm): 210 x 230
Supplier: Chemglass Life Sciences CLS202172
Flask, 2800mL, Fernbach, 70mm Screw Thread, Without Baffles, Vented Cap, Approx OD x Height (mm): 210 x 230. 2800mL flasks are designed for culturing organisms that require a large surface area to volume ratio.