Learn More
BUCHI FlashPure Chromatography Cartride Sample Packs
FlashPure cartridges are offered in a wide range of sizes, covering different stationary phases, particle sizes and geometries for your purification needs.
Supplier: BUCHI 34004988
This item is not returnable.
View return policy
Description
FlashPure EcoFlex cartridges are designed with optimized backpressure, price and performance.
- Normal Phase Bundle for basic applications and polar compounds
- Reverse Phase C18 Bundle for nonpolar compounds and ideal for peptides
- Scale-up Bundle for larger loading separations
Specifications
Silica | |
Sample Bundle of Cartridges | |
Stationary Phase |
Chromatography Cartridge | |
4 g, 12 g, 25 g, 40 g, 80 g |