Learn More
Ace Glass, Inc. Flange style vacuum trap 45mm OD x 280mm l, #50 clamp and O-ring
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ace Glass, Inc. 875513
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
45mm O.D. x 280mm long vacuum trap assembly with o-ring flange connection. Includes #50 stainless steel pinch clamp and Viton o-ring. This design has flexibility and ease of assembly/disassembly for cleaning.