Learn More
AdipoGen FK-506 Tacrolimus
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: AdipoGen AGCN20047M001
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Chemical. CAS 109581-93-3. Formula C44H69NO12 . H2O. MW 804.0 . 18.0. Isolated from Streptomyces FCZ0311. Potent immunosuppressant as cyclosporin A and rapamycin. Suppresses proliferation of cytotoxic T cells and inhibits the production of T cell-derived mediators such as interleukin-2 IL-2. Forms a complex with FK-506 binding protein 12 FKBP12. Inhibits the activity of the calcium/calmodulin-dependent protein phosphatase 2B PP2B calcineurin, leading to disruption of T cell activation. Prevents rejection of transplanted organs. Anti-inflammatory compound in the treatment of several inflammatory skin diseases e.g. atopic dermatitis and with potential anti-rheumatic activity rheumatoid arthritis. Neuroprotective. Regulates nitric oxide neurotoxicity. Neurotransmitter. Ca2 release compound. NF-kappaB suppressor by induction of unfolded protein response UPR. Anti-cancer compound. Apoptosis inducer.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.