Learn More
TARGETMOL CHEMICALS INC FK-453 5MG
Supplier: TARGETMOL CHEMICALS INC T27323
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. FK-453 is a potent antagonist of non-xanthine adenosine A1 receptor with diuretic and renal vasodilatory activity. purity: 99%