Learn More
World Precision Instrument Fixing Screw for connecting INC and GC Neuroscience Cannulas
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: World Precision Instrument 504281
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
- Fixing screw for securing the internal cannula to the guide cannula, to prevent movement during injection
- One size fits all
- We offer a large variety of neuroscience cannulas with many length options. To place an order, Click here for the custom order form and price list, and one of our sales specialists will contact you to complete the order.
Download the catalog page