Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BINDER™ Shelf, Reinforced
Shelf, reinforced - positioned at bottom level, max. load 45 kg, with additional attachment for operation of shaking device, stirring device or roller bottle system
Supplier: BINDER™ 80120288
Description
- Reinforced
- Positioned at bottom level
- Maximum load of 45 kg
Specifications
Shelf, reinforced - positioned at bottom level, max. load 45 kg, with additional attachment for operation of shaking device, stirring device or roller bottle system | |
BINDER Chambers KB 115 | |
Max load of 67 kg | |
Reinforced Shelf |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction