Learn More
Fisher Healthcare™ Sure-Vue™ ASO Test Kit
Increased sensitivity ideal for testing pediatric patients
Specifications
CPT Code 86060, 86063
Type Sure-Vue™ ASO Test Kit
For Use With (Application) To screen both for levels of ASO in Serum greater than 200 IU/mL and levels of ASO greater than 100 IU/mL using an alternative sampling and determine titers of ASO in Serum using the semi-quantitative technique
Test Time 2 min.
View More Specs
Includes: Slides (six-well), latex reagent, positive and negative controls, and package insert
Products 1
Catalog Number Clia Complexity Quantity Includes Price Quantity & Availability  
Catalog Number Clia Complexity Quantity Includes Price Quantity & Availability  
23-038000
View Documents
Moderate 50 Tests Reagent, disposable slides and prediluted positive and negative control Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the supplier.
View Alternate Product
N/A
Description
Description
  • CLIA Complexity: Non-Waived
  • CPT Codes: 86060/86063
  • Two-minute latex agglutination slide test
  • For the qualitative and semi-quantitative determination of antistreptolysin-O in serum
  • Can be used to screen both for levels of ASO in serum greater than 200 IU/mL and levels of ASO greater than 100 IU/mL using an alternative sampling
  • Can also be used to determine titers of ASO in serum using the semi-quantitative technique
  • For in vitro diagnostic use
Specifications
Specifications
86060, 86063
Sure-Vue™ ASO Test Kit
To screen both for levels of ASO in Serum greater than 200 IU/mL and levels of ASO greater than 100 IU/mL using an alternative sampling and determine titers of ASO in Serum using the semi-quantitative technique
2 min.