Learn More
Terra™ Findlay CT FP SD 6 in. Black Boot
The Findlay delivers all around protection with an increased focus on flexibility and range of motion. It provides dry, breathable comfort with its waterproof construction, full-grain waterproof nubuck leather upper, and moisture-wicking, abrasion-resistant liner.
Supplier: Terra™ R5205B 120 M
Description
- Full-grain waterproof nubuck leather upper
- Moisture-wicking and abrasion-resistant aero-lining
- Waterproof boot construction, with CLEANFEET™ odor control treatment
- High-traction, slip-resistant rubber outsole
- Shock absorbing PU midsole and PU toe bumper
- Premium single-density PU FOOTBED™
- CSA Grade 1 composite toe, composite plate, static dissipative
Specifications
Black | |
12 | |
Yes | |
Work Boot |
Leather | |
Men's | |
Yes |