Learn More
Perkin Elmer US LLC FID Gas Station - NM Plus, 350 mL/min H2 and 1500 mL/min Air
Supplier: Perkin Elmer US LLC N9300016
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
Designed with space saving in mind, it sits directly under the GC to conserve bench space. You can also apply the FID gas station to GCMS systems with the simple addition of a drawer tray for the MS to rest on. If you want to benefit from the space saving design, but only require a H2 generator, that is also an option. Air Flow Rate: 1500 mL/min H2 Flow Rate: 350 mL/min Zero Air Flow Rate: 5 L/min Zero Air Purity: <0.1 ppm Certifications: CE, CSA, FCC
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.