Learn More
Sigma Aldrich Fine Chemicals Biosciences Fibronectin bovine plasma, 1 mg
Supplier: Sigma Aldrich Fine Chemicals Biosciences F11411MG
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Fibronectin is a ubiquitous protein of the extracellular matrix. It is a mosaic protein, that is made up of three types of repeating modular consensus amino acid sequences. Fibronectins belongs to a class of high-molecular-weight glycoproteins. Cellular fibronectin is expressed by fibroblasts and plasma fibronectin is usually liberated by hepatocytes.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.