Learn More
Alpco Ltd Fibrinogen ELISA
SDP
Supplier:  Alpco Ltd 41FIBHUE01
Encompass
The Fibrinogen ELISA is for the determination of fibrinogen in human Serum and other Biological samples. Fibrinogen contributes to the formation of fibrin clots. As an acute phase protein, high levels of fibrinogen may be associated with cardiovascular disease (especially thrombosis). Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC2885199