Learn More
Jackson Safety Face Shield Cap Adapter
The Jackson Safety 282 B Cord Mounted Cap Adapters for Non-Slotted Caps features a polyethylene spark deflector and is ideal for when the job requires a hard hat and a face shield.
Supplier: Jackson Safety 14945
This item is not returnable.
View return policy
Description
- Comes in 3 Models: Model 37, 137, and 337
- Curved injection molded channel fit over the hard hat brim (Model 337)
- Features adjustable rubber strap headband that stretches around the cap crown for a secure fit (Model 337)
- Features five easy-to-swivel lock mounting pins for universal windows
- Vertical lock-in eliminates face shield creep (Model 37, 137)
- Curved metal channels fit over the hard hat brim
- Brackets feature durable expandable spring tensioner (Model 37, 137)Features five easy-to-swivel lock mounting pins for universal windows
Specifications
Silver | |
Face Shield Cap Adapter | |
Spring | |
Universal |
Face Shields: Full Brim Hard Hats | |
Metal | |
Face Shield Cap Adapter Bracket | |
Full Brim |