Learn More
Glass Expansion EzyLok Connector 4mm Tubing
Supplier: Glass Expansion QSM4
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
EzyLok Connector 4mm Tubing. The EzyLok connector is an easy, safe, and effective way to connect argon supply tubes to your Glass Expansion F-Series and U-Series nebulizers. Can be used with 4mm OD nylon tubing, or standard 6mm OD (1/4” OD, 1/8” ID) soft walled tubing when using the EzyLok 6mm Hose Adaptor (70-808-0735). The EzyLok connection is made with Glass Expansion’s specially formed nebulizer side arm, which allows it to be pushed and locked securely into the popular “One Touch Fitting”, that incorporates a flexible O-Ring seal. To disconnect the nebulizer, simply push on the release button located at the top of the fitting.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.