Learn More
Glass Expansion EzyFit Sample Tube - Twin (PKT 6)
Supplier: Glass Expansion EZT1650T
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
EzyFit Sample Tube - Twin (PKT 6). A replacement 1.6mm (1/16”) OD and 0.50mm ID capillary with an EzyFit connector on both ends. Typically used with Glass Expansion’s Trident IS Kit P/N NC9281090, 60-808-1185. This fitting connects the mixed sample and internal standard from the mixing T to a Glass Expansion F-Series nebulizer.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.