Learn More
Glass Expansion EzyFit with 1.3mm OD x 0.75mm ID x 1500mm long sample tube (PKT. 10)
Supplier: Glass Expansion EZTS075
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
EzyFit with 1.3mm OD X 0.75mm ID x 1500mm long sample tube. Package of 10 replacement sample lines compatible with Glass Expansion F-Series Nebulizers. Used to carry sample from peristaltic pump tubing or mixing T to nebulizer. This consumable part should be replaced at a frequency of about every 6 months to ensure optimum sample flow.For EzyFit Connector with 0.50mm ID, 1/16 OD x 700mm long sample tube, EZT-16-50. For EzyFit Connector with 0.25mm ID, 1.3mm OD x 700mm long sample tube, EZT-025.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.