Learn More
Apexbio Technology LLC EZLink Sulfo-NHS-Biotin Kit, 10 reactions
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Apexbio Technology LLC K100510
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The EZLink Sulfo-NHS-Biotin Kit contains reagents sufficient for 10 biotin labeling reactions. It is a water-soluble biotin reagent for labeling protein, antibody or other molecules with amidogen (NH2-). The biotin in the kit is sufficient and has been activated for direct use. The whole procedure is simple and quick. Other sizes are available. Please inqury us for quote.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.