Learn More
Medtox™ Diagnostics EZ-SCREEN™ (HI) 4 Panel
Ideal for standard drugs of abuse testing in workplaces, EZ-SCREEN (HI) delivers fast, accurate screening results in minutes for comprehensive drugs of abuse testing that comes complete with an on-board adulteration strip.
Supplier: Medtox™ Diagnostics 21500134
Description
- Screens for Amphetamine, Cocaine, Opiates (2000), PCP
- Includes adulteration strip
- 510(k) cleared
- Quality management system conforms with ISO 13485
- Accurate screening results in minutes
- Made in the USA
Specifications
No | |
Amphetamine, Cocaine (Benzoylecgonine), Opiates | |
Drug Panel (Urine) |
Moderate Complexity | |
Single-use |