Learn More
Market Lab Eye Shields, 10pk
Supplier: Market Lab 7046
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The MarketLab Face Shield is a clear plastic face shield that provides protection for the face and eyes against splashes, sprays, and respiratory droplets. It has an adjustable headgear with foam padding for comfort and can be disinfected and reused. The shield measures 8" x 12" and the headgear adjusts from 6.5" to 8". It is made from optically clear PETG plastic for high visibility. The Face Shield can be worn with other PPE like masks and goggles. It is lightweight, comfortable and provides full face coverage.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.