Learn More
Electron Microscopy Sciences ExP Vacuum Wand Kit for 6" (150mm) wafers, Molded PEEK wafer tip, ⅛" coiled hose and stand
Supplier: Electron Microscopy Sciences 7193201
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
ExP Vacuum Wand wafer handling kits are a lower cost alternative to the Porta-Wand and Porta-Vac II systems, which share the same tips. Has a push-button to close the vacuum and release the wafer. The included stand keeps the button pressed when the wand is not in use. Comes complete with an ⅛" Inner Diameter coiled vacuum hose and a wafer tip. Must use a vacuum pump or vacuum line. Able to be used with our Vacuum Pick Up System. Comes with 1/8" coiled hose and stand.
Includes: Molded PEEK wafer tip, ⅛" coiled hose and stand
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.