Learn More
Medchemexpress LLC HY-17613 5mg Medchemexpress, Evocalcet CAS:870964-67-3 Purity:>98%
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Medchemexpress LLC HY176135MG
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Medchemexpress, HY-17613 5mg Evocalcet CAS:870964-67-3 Evocalcet has an activating effect on calcium sensing receptor (CaSR) extracted from patent WO 2017061621 A1, compound A. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.