Learn More
Selleck Chemical LLC Evocalcet-5mg
Supplier: Selleck Chemical LLC E0359
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Evocalcet (KHK7580) is an novel oral calcimimetic agent which acts via allosteric activation of the calcium sensing receptor (CaSR). Evocalcet (0, 20 and 60 nM) dose-dependently increases cytoplasmic [Ca2+] concentrations with an EC50 value of 92.7 nM in hCaR-HEK293 cells.