Learn More
Glas Col LLC Evaporator
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Glas Col LLC 109A 1180485
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Evaporator for (48) 12mm vials insert/gas inlet heated to maximum 100 degrees C, 120V