Learn More
Glas Col LLC Evaporator
SDP
Supplier:  Glas Col LLC 109A 1180333CE
Encompass_Preferred
Request Bulk Order
Evaporator 96 well plates (3) insert and gas inlet heated to 100 degrees C. 240V CE
Catalog No. NC1815886