Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ ETP 45835 dihydrochloride
Mnk1 and Mnk2 inhibitor
$299.28 - $1212.20
Specifications
Purity | >99% |
---|---|
Product Type | ETP 45835 dihydrochloride |
Molecular Weight (g/mol) | 301.21 |
Inhibitors | ETP 45835 dihydrochloride |
Description
ETP 45835 dihydrochloride is a Mnk2 and Mnk1 inhibitor (IC50 values are 575 and 646 nM respectively). Exhibits selectivity against a panel of 24 protein kinases, including those upstream of Mnk1/2.Specifications
>99% | |
ETP 45835 dihydrochloride | |
301.21 | |
ETP 45835 dihydrochloride |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title