Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-methyl-1,2,3-thiadiazole-5-carboxylate, 98%, Thermo Scientific Chemicals

Catalog No. AAA1339906
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18212-20-9
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.20
MDL Number MFCD00052549
InChI Key AHPXTXGCMLOXGA-UHFFFAOYSA-N
Synonym ethyl 4-methyl-1,2,3-thiadiazole-5-carboxylate, 4-methyl-1,2,3-thiadiazole-5-carboxylic acid ethylester, 1,2,3-thiadiazole-5-carboxylic acid, 4-methyl-, ethyl ester, acmc-209xfp, ethylmethylthiadiazolecarboxylate, 5-ethoxycarbonyl-4-methyl-1,2,3-thiadiazole, 4-methyl-5-thiadiazolecarboxylic acid ethyl ester, 4-methyl-1,2,3 thiadiazole-5-carboxylic acid ethyl ester, 4-methyl-1,2,3-thiadiazole-5-carboxylic acid ethyl ester
PubChem CID 2758878
IUPAC Name ethyl 4-methylthiadiazole-5-carboxylate
SMILES CCOC(=O)C1=C(C)N=NS1

Specifications

Density 1.265
Boiling Point 117°C to 118°C (15 mmHg)
Refractive Index 1.504
Quantity 5 g
Beilstein 609140
Formula Weight 172.21
Percent Purity 98%
Chemical Name or Material Ethyl 4-methyl-1,2,3-thiadiazole-5-carboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.