Learn More
PREDICTIVE ONCOLOGY INC Endotoxin Sample Prepration (ESP) Kit
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: PREDICTIVE ONCOLOGY INC ESP9001.01
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
ESP™ is a sample treatment system to remove the inhibitory or enhancing effects of blood products in endotoxin assays. This allows for more accurate endotoxin detection and quantitation. The ESP™ system is compatible with both classical LAL and recombinant Factor C endotoxin detection assays.