Learn More
Electron Microscopy Sciences EMS Carbon Rods
Supplier: Electron Microscopy Sciences 91077
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Carbon Rods for the EMS 975 and 975S turbo evaporator. Rods have a 6.15mm diameter.