Learn More
MilliporeSigma™ Chemicon™ Caspase 9 Colorimetric Assay Kit, LEHD
Provide a simple and convenient means for assaying the activity of caspases that recognize the sequence LEHD
Supplier: MilliporeSigma™ APT173
Specifications
-20°C | |
Chromogenic | |
pNA Standard, 5X Cell Lysis Buffer, 5X Assay Buffer, Caspase-9 Substrate, Caspase-9 Inhibitor |
Caspase 9 Colorimetric Assay Kit, LEHD | |
Provide a simple and convenient means for assaying the activity of caspases that recognize the sequence LEHD | |
100 Tests |