Learn More
Newcomer Supply Embedding Tissue Tampers, 16mm x 19mm
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Newcomer Supply 5108B
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Newcomer Supply’s Tissue Embedding Tampers are unique tools for flattening tissue sections in an embedding mold with ease! The design allows for the end-user to either use a forceps or their fingers to easily control the tampers. The aluminum, one piece handle extends 24mm above the tamper base and comes with a vinyl cap to help remain cool to the touch. There is also an anodized colored surface on the tamper that makes it more durable and corrosion-resistant. The tampers comes in two different sizes to accommodate small, medium and large embedding molds: 10mm x 10mm base (orange) and 16mm x19mm base (green). A combo pack is also available including 1 of each size.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.