Learn More
GVS FILTER TECHNOLOGY INC Elipse P100 nuisance odour ready to use Mask with replaceable filters (M/L size)
Supplier: GVS FILTER TECHNOLOGY INC SPR456
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This item has a minimum qty of 3 per supplier requirements.
GVS designed the Elipse respirators to fit the contours of the individuals face & the result is a form fitting respirator users will find very comfortable. Elipse masks also have been designed to completely seal around the nose area to prevent exhaled air from fogging the wearers safety glasses. low profile filter design offers a unique solution to customers by providing the user w/ a lightweight & comfortable respirator. Elipse provides wearers w/ a wide field of view & filters that don't interfere w/ other PPE such as sanding shields, safety glasses or welding hoods. The innovative close pleated composite P-100 filters provide users w/ the maximum particulate protection in a small & lightweight filter. Elipse respirators are re-usable & the intuitive design allows the filters to easily be interchanged. A low breathing resistance , 4 point harness for secure fit & soft thermoplastic elastomer mask material.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.