Learn More
Cole-Parmer Cole-Parmer Single-Junction ORP Electrode, Platinum Sensor, 10-ft Cable 05994-21
Supplier: Cole-Parmer 0599421
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
ORP electrodes measure oxidation/reduction levels in plating baths, waste water monitoring, and more. Use platinum-band electrodes for general-purpose applications from 23 to 212°>F (-5 to 100°>C). Use gold-disc electrodes for solutions containing cyanide from 23 to 230°>F (-5 to 110°>C).