Learn More
Eisco™ Visual Scientifics Loop
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Loop investigation demonstrates conservation of kinetic and potential energy.
Supplier: Eisco™ PTLOP
Includes: Extensive instructions to assist with lesson planning, student activities and experimental procedures; includes metal ramp with loop and two metal balls
Description
Students explore the real physics of roller coaster loops and then run experiments to see if their loops work.
- Main Lab: Is the ball going to make it around the loop? Do roller coasters knock people out?
- Extension One: normal force on a roller coaster rider
- Extension Two: technology verification of calculations
- Covers 15 Next Generation Science Standards Concepts
- Covers 17 Common Core Standards
- Approximately three hours of class/lab time material
- 30 pages in total
- Student workbook is a guide for students to fill out as they work through the main lab and lab extensions
- General Theory Section lays out the theory and mathematics behind the lesson plan
- Experimental Teaching Guide helps teachers prepare lesson plans and lead inquiry or step-based activities
- All labs and extensions include recommendations for video analysis setups
Order Info
For use with Physical Science System Base (S06893).
Specifications
Loop | |
Extensive instructions to assist with lesson planning, student activities and experimental procedures, includes metal ramp with loop and two metal balls | |
Loop investigation demonstrates conservation of kinetic and potential energy | |
Physics |