Learn More
Gold Biotechnology Inc EDTA Disodium, dihydrate 100 g
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Gold Biotechnology Inc E210100
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
EDTA (Ethylenediaminetetraacetic acid) is a chelating agent, a general chemical and a sequestrant. In molecular biology applications, it is used to minimize metal ion contamination and prevent enzymatic activity.