Learn More
Across International ECO 150C 1.9 Cu Ft Vacuum Drying Oven with LED Lights
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Across International AT19E
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
AT19e series digital desktop vacuum ovens feature a 1.9 cubic foot easy-to-clean stainless steel chamber with a large dual-layer tempered glass safety window and small footprint. These ovens can greatly reduce drying time by maintaining a consistent vacuum level within the chamber.
This item is not returnable.
View return policy