Learn More
Mortara Instrument Inc ECG SYSTEM ELI 230 110 V
Supplier: Mortara Instrument Inc BUR230A
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
ECG System ELI™ 230 110 V Color DisplayDelivers impressive 12-lead performance, while requiring minimal space0.05 to 300 Hz Frequency40,000 Sampling rateMemory capacity- 20 patient storageIncludes AM12 USB 20 patient storage
This item is not returnable.
View return policy