Learn More
Merry X Ray Corp E Z Store & Pour Developer 4 x 1 Gal
Supplier: Merry X Ray Corp 103633
E-Z Store & Pour Developer is conveniently packaged in easy to handle one-gallon bottles, four per case. No mixing required.Specifically formulated for long life in low volume situations, sealed gallon bottles of E-Z Store & Pour Developer have a shelf life of up to one year. Once opened, this chemical should be used within 2 weeks to minimize chemical oxidation.Ready-to-use (premixed) developer solution ideal for low volume accounts, i.e., less than 25-14x17" films processed per eight hours.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.